Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199495.2 | NP_955789.2 | 2 | 235 | ||
XM_008767385.1 | XP_008765607.1 | 2 | 247 | |||
GenBank mRNA | BC093375.1 | - | 2 | 194 | 7389276 | |
CB610197.1 | - | 2 | 235 | |||
FQ209525.1 | - | 2 | 237 |
Gene Symbol: Ndufa10l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182671.2 | NP_872612.1 | 1 | 197 | ||
GenBank mRNA | AY260163.1 | - | 1 | 197 |