Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18206
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
10
1532
NM_001258380.1
NP_001245309.1
10
1294
NM_001321103.1
NP_001308032.1
11
1916
NM_001321104.1
NP_001308033.1
11
1877
NM_001321105.1
NP_001308034.1
11
1877
NM_001321106.1
NP_001308035.1
11
1865
NM_001321107.1
NP_001308036.1
10
1532
NM_001321108.1
NP_001308037.1
10
1544
NM_003615.4
NP_003606.3
11
1666
NR_135541.1
11
1865
NR_135542.1
10
1532
NR_135543.1
10
1532
NR_135544.1
11
1865
XM_005265598.4
XP_005265655.1
10
1798
XM_005265600.4
XP_005265657.1
10
1760
XM_006713421.2
XP_006713484.1
11
1904
XM_011534256.2
XP_011532558.1
11
2177
XM_011534258.1
XP_011532560.1
11
1880
XM_011534261.2
XP_011532563.1
11
2142
XM_011534263.2
XP_011532565.1
10
1469
XM_011534265.1
XP_011532567.1
10
1508
XM_017007526.1
XP_016863015.1
12
1801
XM_017007527.1
XP_016863016.1
10
1493
XM_017007528.1
XP_016863017.1
4
417
XM_017007529.1
XP_016863018.1
4
399
GenBank mRNA
AB012130.1
-
7
980
AF047033.1
-
11
1666
AF053755.1
-
7
972
AF089726.1
-
10
1294
CR627428.1
-
3
248
EU499349.1
-
10
1459
EU934246.1
-
11
1792
EU934247.1
-
11
1804
EU934248.1
-
11
1843
EU934249.1
-
10
1459
EU934250.1
-
11
1804
FJ178574.1
-
11
1792
FJ178575.1
-
10
1459
FJ178576.1
-
10
1459
GU354307.1
-
11
1792
GU354308.1
-
10
1471
GU354309.1
-
10
1471
GU354310.1
-
10
1471
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0