Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VPS4B SIRNA Assays ›
          Gene Symbol:
          VPS4B
          Gene Name:
          vacuolar protein sorting 4 homolog B
          Gene Aliases:
          MIG1, SKD1, SKD1B, VPS4-2
          Chromosome Location:
          Chr.18: 63389190 - 63422519 on Build GRCh38
          UniGene ID:
          Hs.126550
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18272

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VPS4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004869.3 NP_004860.2 7 1020
          XM_006722582.2 XP_006722645.1 6 896
          XM_011526268.2 XP_011524570.1 7 1020
          GenBank mRNA AF038960.1 - 7 949
          AF195514.1 - 7 961
          AK290932.1 - 7 964
          AK291253.1 - 7 918
          AK308892.1 - 7 984
          AK309322.1 - 6 896
          AK311041.1 - 6 896
          AY232629.1 - 7 770
          BC039574.1 - 7 927 6042862

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell separation after cytokinesis
          potassium ion transport
          autophagy
          nucleus organization
          vacuole organization
          mitotic metaphase plate congression
          regulation of centrosome duplication
          positive regulation of G2/M transition of mitotic cell cycle
          protein transport
          endosomal transport
          viral life cycle
          viral release from host cell
          cholesterol transport
          cytoplasmic microtubule organization
          endosome to lysosome transport via multivesicular body sorting pathway
          response to lipid
          multivesicular body assembly
          viral budding via host ESCRT complex
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          positive regulation of viral process
          regulation of viral process
          protein depolymerization
          negative regulation of cell death
          late endosomal microautophagy
          ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway
          regulation of mitotic spindle assembly
          positive regulation of viral release from host cell
          negative regulation of exosomal secretion
          positive regulation of exosomal secretion
          positive regulation of centriole elongation
          positive regulation of viral life cycle
          ESCRT III complex disassembly
          protein binding
          ATP binding
          protein C-terminus binding
          microtubule-severing ATPase activity
          ATPase activity
          ATPase activity, coupled
          identical protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0