Search
Search

| Gene Symbol: Tceal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577026.6 | XP_577026.4 | 3 | 368 | ||
| GenBank mRNA | FQ213749.1 | - | 3 | 306 | ||
| Gene Symbol: Tceal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749245.3 | XP_003749293.1 | 3 | 380 | ||
| GenBank mRNA | FQ213604.1 | - | 3 | 306 | ||