siRNA Details
Gene Symbol: AUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306190.1 | NP_001293119.1 | 7, 8 | 892 | ||
NM_001698.2 | NP_001689.1 | 8, 9 | 934 | |||
XM_005252066.3 | XP_005252123.1 | 9, 10 | 977 | |||
XM_006717150.3 | XP_006717213.1 | 8, 9 | 893 | |||
XM_011518802.2 | XP_011517104.1 | 9, 10 | 1003 | |||
XM_017014850.1 | XP_016870339.1 | 9, 10 | 1074 | |||
GenBank mRNA | BC020722.1 | - | 7, 8 | 836 | 4656960 | |
X79888.1 | - | 8, 9 | 903 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Molecular Function -










