Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGDCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004884.3 | NP_004875.2 | 3 | 787 | ||
XM_011522241.1 | XP_011520543.1 | 3 | 729 | |||
XM_011522243.1 | XP_011520545.1 | 2 | 313 | |||
XM_011522244.1 | XP_011520546.1 | 2 | 302 | |||
XM_011522245.2 | XP_011520547.1 | 3 | 729 | |||
GenBank mRNA | AK095529.1 | - | 2 | 302 | ||
BC042054.1 | - | 3 | 735 | 5579570 | ||
BC067107.1 | - | 3 | 787 | 6644528 |