siRNA Details
Gene Symbol: RGD1566008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257584.2 | XP_006257646.1 | 1 | 236 |

Gene Symbol: RGD1566008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257584.2 | XP_006257646.1 | 1 | 236 |