Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNCAIP SIRNA Assays ›
Gene Symbol:
SNCAIP
Gene Name:
synuclein alpha interacting protein
Gene Aliases:
SYPH1, Sph1
Chromosome Location:
Chr.5: 122311354 - 122464219 on Build GRCh38
UniGene ID:
Hs.426463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18501
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNCAIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242935.2
NP_001229864.1
3
366
NM_001308100.1
NP_001295029.1
5
1811
NM_001308105.1
NP_001295034.1
2
150
NM_001308106.1
NP_001295035.1
3
291
NM_001308107.1
NP_001295036.1
3
366
NM_001308108.1
NP_001295037.1
2
150
NM_001308109.1
NP_001295038.1
3
366
NM_005460.3
NP_005451.2
3
366
NR_131761.1
3
291
NR_131762.1
3
242
XM_005272138.3
XP_005272195.1
3
289
XM_005272139.1
XP_005272196.1
3
229
XM_006714734.2
XP_006714797.1
3
422
XM_011543737.2
XP_011542039.1
4
424
XM_011543738.2
XP_011542040.1
4
423
XM_011543739.1
XP_011542041.1
4
349
XM_011543741.2
XP_011542043.1
4
373
XM_011543743.2
XP_011542045.1
4
798
XM_011543745.1
XP_011542047.1
3
253
XM_011543749.2
XP_011542051.1
3
656
XM_011543750.1
XP_011542052.1
2
134
XM_017010078.1
XP_016865567.1
4
2751
XM_017010079.1
XP_016865568.1
4
424
XM_017010080.1
XP_016865569.1
4
424
XM_017010081.1
XP_016865570.1
4
422
XM_017010082.1
XP_016865571.1
3
253
XM_017010083.1
XP_016865572.1
3
282
XM_017010084.1
XP_016865573.1
4
430
XM_017010085.1
XP_016865574.1
4
361
XM_017010086.1
XP_016865575.1
4
428
GenBank mRNA
AB110788.1
-
2
104
AB110789.1
-
2
104
AB110790.1
-
3
245
AB110791.1
-
3
245
AB110792.1
-
2
104
AB110793.1
-
2
104
AB110794.1
-
2
104
AF076929.1
-
3
197
AK298882.1
-
3
247
AK299687.1
-
3
242
AK304646.1
-
3
198
AK310835.1
-
3
198
BC033743.1
-
3
186
5193089
BC040552.1
-
5
1673
5298588
BC094759.1
-
3
244
4838145
BM908442.1
-
310
DQ227317.1
-
3
310
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell death
dopamine metabolic process
cellular protein metabolic process
regulation of neurotransmitter secretion
regulation of inclusion body assembly
protein binding
ubiquitin protein ligase binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0