Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2M
          Gene Name:
          beta-2-microglobulin
          Gene Aliases:
          IMD43
          Chromosome Location:
          Chr.15: 44711487 - 44718159 on Build GRCh38
          UniGene ID:
          Hs.534255
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1853

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2M
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004048.2 NP_004039.1 1, 2 131
          XM_005254549.3 XP_005254606.1 1, 2 126
          GenBank mRNA AB021288.1 - 1, 2 84
          AF072097.1 - 871
          AK315776.1 - 1, 2 101
          AY007153.1 - 1, 2 83
          AY187687.1 - 1, 2 71
          BC032589.1 - 1, 2 105 5502428
          BC064910.1 - 1, 2 88 6055473
          BU658737.1 - 1, 2 143
          CR457066.1 - 1, 2 71
          DQ839493.1 - 1, 2 71
          DQ839494.1 - 1, 2 71
          DQ884406.1 - 1, 2 71
          S54761.1 - 1, 2 96
          S82300.1 - 1, 2 102
          V00567.1 - 1, 2 44
          X07621.1 - 1, 2 86

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          positive regulation of T cell cytokine production
          regulation of membrane depolarization
          negative regulation of neuron projection development
          antibacterial humoral response
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          positive regulation of protein binding
          T cell differentiation in thymus
          protein refolding
          response to drug
          cellular protein metabolic process
          innate immune response
          response to cadmium ion
          positive regulation of receptor-mediated endocytosis
          regulation of defense response to virus by virus
          regulation of immune response
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          iron ion homeostasis
          interferon-gamma-mediated signaling pathway
          cellular response to lipopolysaccharide
          cellular response to iron ion
          negative regulation of receptor binding
          positive regulation of receptor binding
          positive regulation of ferrous iron import into cell
          positive regulation of ferrous iron binding
          positive regulation of transferrin receptor binding
          glycoprotein binding
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0