Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K1 SIRNA Assays ›
Gene Symbol:
PPIP5K1
Gene Name:
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases:
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location:
Chr.15: 43533462 - 43590272 on Build GRCh38
UniGene ID:
Hs.156814
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18624
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130858.2
NP_001124330.1
19
2244
NM_001130859.2
NP_001124331.1
18
2239
NM_001190214.1
NP_001177143.1
19
2244
NM_014659.5
NP_055474.3
18
2162
XM_005254803.1
XP_005254860.1
19
2244
XM_005254804.1
XP_005254861.1
19
2244
XM_006720786.2
XP_006720849.1
19
2244
XM_011522249.1
XP_011520551.1
19
2244
XM_011522250.1
XP_011520552.1
19
2347
XM_011522252.1
XP_011520554.1
18
2218
XM_011522254.1
XP_011520556.1
18
2114
XM_011522258.1
XP_011520560.1
13
1504
XM_011522259.1
XP_011520561.1
19
2244
XM_017022735.1
XP_016878224.1
18
2561
XM_017022736.1
XP_016878225.1
18
2529
XM_017022737.1
XP_016878226.1
18
2210
XM_017022738.1
XP_016878227.1
18
2158
XM_017022739.1
XP_016878228.1
18
2158
XM_017022740.1
XP_016878229.1
19
2347
XM_017022741.1
XP_016878230.1
18
2560
XM_017022742.1
XP_016878231.1
19
2352
XM_017022743.1
XP_016878232.1
18
2218
XM_017022744.1
XP_016878233.1
18
2159
XM_017022745.1
XP_016878234.1
19
2347
XM_017022746.1
XP_016878235.1
18
2218
XM_017022747.1
XP_016878236.1
18
2158
XM_017022748.1
XP_016878237.1
18
2560
XM_017022749.1
XP_016878238.1
19
2320
XM_017022750.1
XP_016878239.1
18
2161
XM_017022751.1
XP_016878240.1
18
2114
XM_017022752.1
XP_016878241.1
19
2244
XM_017022753.1
XP_016878242.1
18
2218
XM_017022754.1
XP_016878243.1
19
2352
XM_017022755.1
XP_016878244.1
18
2218
XM_017022756.1
XP_016878245.1
18
2156
XM_017022757.1
XP_016878246.1
18
2114
XM_017022758.1
XP_016878247.1
13
1504
XM_017022759.1
XP_016878248.1
18
2158
XM_017022760.1
XP_016878249.1
18
3692
GenBank mRNA
AB002375.1
-
18
2166
AF502586.1
-
18
2134
AF502587.1
-
18
2166
AF502588.1
-
18
2134
AF502589.1
-
18
2166
AF543190.1
-
18
2116
AK307438.1
-
19
2208
BC050263.1
-
17
2190
5521199
BC057395.1
-
18
2190
6142663
BX538022.1
-
18
2190
BX647814.1
-
19
2196
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0