siRNA Details
Gene Symbol: LOC102554287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257513.1 | XP_006257575.1 | 1 | 209 |

Gene Symbol: LOC102554287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257513.1 | XP_006257575.1 | 1 | 209 |