Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000114.1 | NP_001000114.1 | 1 | 536 |
Gene Symbol: LOC684403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770643.1 | XP_008768865.1 | 1 | 536 |
Gene Symbol: Olr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000115.1 | NP_001000115.1 | 1 | 536 |