Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077683.2 | NP_001071151.2 | 4 | 2070 | ||
XM_006240664.2 | XP_006240726.1 | 5 | 2224 |