Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304743.1 | NP_001291672.1 | 12 | 3197 | ||
NM_014720.3 | NP_055535.2 | 12 | 3197 | |||
XM_011540401.2 | XP_011538703.1 | 11 | 1853 | |||
GenBank mRNA | AB002804.1 | - | 12 | 2868 | ||
AF273048.1 | - | 12 | 2670 | |||
AK302855.1 | - | 12 | 2630 | |||
BC111565.1 | - | 12 | 2667 | 40009070 | ||
D86959.1 | - | 12 | 3147 |