Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235641.1 | XP_006235703.1 | 14 | 6326 | ||
XM_006235642.2 | XP_006235704.1 | 14 | 6066 | |||
XM_006235643.2 | XP_006235705.1 | 14 | 5974 |