Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135913.1 | NP_001129385.1 | 7 | 1774 | ||
XM_006242122.2 | XP_006242184.1 | 8 | 2271 | |||
XM_008765787.1 | XP_008764009.1 | 8 | 2304 | |||
XM_008765788.1 | XP_008764010.1 | 8 | 2271 | |||
XM_008765789.1 | XP_008764011.1 | 6 | 1490 | |||
XM_008765790.1 | XP_008764012.1 | 6 | 2169 | |||
XM_008765791.1 | XP_008764013.1 | 6 | 1454 | |||
GenBank mRNA | BC167764.1 | - | 7 | 1774 | 7453894 |