Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013209.1 | NP_001013227.1 | 5 | 448 | ||
XM_006235863.2 | XP_006235925.1 | 5 | 494 | |||
XM_006235864.2 | XP_006235926.1 | 6 | 551 | |||
XM_006235866.2 | XP_006235928.1 | 5 | 491 | |||
GenBank mRNA | BC078908.1 | - | 5 | 448 | 7113862 | |
FQ214601.1 | - | 5 | 449 |
Gene Symbol: LOC690183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001073579.4 | XP_001073579.1 | 1 | 442 |