Search
Search

CEP350| Gene Symbol: CEP350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014810.4 | NP_055625.4 | 34 | 8136 | ||
| XM_011510191.2 | XP_011508493.1 | 36 | 8311 | |||
| XM_011510198.2 | XP_011508500.1 | 35 | 8188 | |||
| XM_017002929.1 | XP_016858418.1 | 35 | 8245 | |||
| XM_017002930.1 | XP_016858419.1 | 35 | 8283 | |||
| XM_017002931.1 | XP_016858420.1 | 35 | 8215 | |||
| GenBank mRNA | AB007949.1 | - | 8 | 2291 | ||
| AF287356.1 | - | 34 | 7920 | |||
| AK022813.1 | - | 1 | 145 | |||