Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750867.3 | XP_003750915.1 | 2 | 991 |
Gene Symbol: RGD1564405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749882.3 | XP_003749930.1 | 2 | 822 |