Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MED24 SIRNA Assays ›
          Gene Symbol:
          MED24
          Gene Name:
          mediator complex subunit 24
          Gene Aliases:
          ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
          Chromosome Location:
          Chr.17: 40019097 - 40054636 on Build GRCh38
          UniGene ID:
          Hs.462983
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19096

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MED24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079518.1 NP_001072986.1 19, 20 2538
          NM_001267797.1 NP_001254726.1 19, 20 2516
          NM_014815.3 NP_055630.2 20, 21 2555
          NR_052017.1 19, 20 2481
          XM_005257874.2 XP_005257931.1 20, 21 2460
          XM_006722204.2 XP_006722267.1 20, 21 2496
          XM_006722206.2 XP_006722269.1 20, 21 2404
          XM_006722207.2 XP_006722270.1 20, 21 2372
          XM_011525529.2 XP_011523831.1 21, 22 2553
          XM_011525530.2 XP_011523832.1 21, 22 2393
          XM_011525531.2 XP_011523833.1 20, 21 2336
          XM_011525532.2 XP_011523834.1 21, 22 2461
          XM_011525533.2 XP_011523835.1 21, 22 2429
          XM_011525534.2 XP_011523836.1 20, 21 2372
          XM_011525535.2 XP_011523837.1 20, 21 2372
          XM_011525536.2 XP_011523838.1 21, 22 2393
          XM_011525538.2 XP_011523840.1 20, 21 2687
          XM_017025458.1 XP_016880947.1 21, 22 2425
          XM_017025459.1 XP_016880948.1 20, 21 2368
          XM_017025460.1 XP_016880949.1 21, 22 2425
          XM_017025461.1 XP_016880950.1 21, 22 2517
          XM_017025462.1 XP_016880951.1 21, 22 2425
          XM_017025463.1 XP_016880952.1 21, 22 2393
          XM_017025464.1 XP_016880953.1 20, 21 2372
          XM_017025465.1 XP_016880954.1 20, 21 2496
          XM_017025466.1 XP_016880955.1 20, 21 2368
          XM_017025467.1 XP_016880956.1 20, 21 2368
          XM_017025468.1 XP_016880957.1 20, 21 2336
          XM_017025469.1 XP_016880958.1 20, 21 2651
          XM_017025470.1 XP_016880959.1 20, 21 2651
          GenBank mRNA AF055995.1 - 19, 20 2242
          AF277379.1 - 19, 20 2242
          AK022508.1 - 19, 20 2307
          AK291040.1 - 19, 20 2287
          AK299864.1 - 20, 21 2172
          AK300992.1 - 19, 20 2160
          AK303502.1 - 8, 9 1010
          AK303745.1 - 19, 20 2251
          BC011375.1 - 19, 20 2517 3866038
          D50920.1 - 20, 21 2315
          EU392533.1 - 1, 2 22
          EU392537.1 - 1, 2 22
          EU392538.1 - 1, 2 22
          EU392540.1 - 7, 8 870
          EU392542.1 - 5, 6 600

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001034079, XM_006247416, XM_006247415, BC097461.
          This siRNA also targets the following transcripts in Mouse: NM_011869, BC078459, AF126543, AF483499, AF483498, and others.

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          protein ubiquitination
          histone acetylation
          stem cell population maintenance
          intracellular steroid hormone receptor signaling pathway
          androgen receptor signaling pathway
          positive regulation of transcription, DNA-templated
          protein heterooligomerization
          RNA polymerase II transcription cofactor activity
          transcription cofactor activity
          histone acetyltransferase activity
          receptor activity
          protein binding
          ligand-dependent nuclear receptor transcription coactivator activity
          vitamin D receptor binding
          thyroid hormone receptor binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0