Search
Search

MAGI2| Gene Symbol: MAGI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301128.1 | NP_001288057.1 | 4 | 899 | ||
| NM_012301.3 | NP_036433.2 | 4 | 899 | |||
| XM_011516718.1 | XP_011515020.1 | 4 | 1130 | |||
| XM_011516719.2 | XP_011515021.1 | 3 | 4418 | |||
| XM_011516720.2 | XP_011515022.1 | 3 | 9408 | |||
| XM_017012840.1 | XP_016868329.1 | 4 | 1130 | |||
| XM_017012841.1 | XP_016868330.1 | 4 | 1130 | |||
| XM_017012842.1 | XP_016868331.1 | 4 | 1130 | |||
| XM_017012843.1 | XP_016868332.1 | 4 | 1130 | |||
| XM_017012844.1 | XP_016868333.1 | 4 | 1130 | |||
| XM_017012845.1 | XP_016868334.1 | 4 | 1130 | |||
| XM_017012846.1 | XP_016868335.1 | 4 | 1130 | |||
| XM_017012847.1 | XP_016868336.1 | 4 | 7367 | |||
| XM_017012848.1 | XP_016868337.1 | 3 | 4415 | |||
| XM_017012849.1 | XP_016868338.1 | 3 | 4415 | |||
| XM_017012850.1 | XP_016868339.1 | 4 | 1130 | |||
| XM_017012851.1 | XP_016868340.1 | 4 | 1130 | |||
| XM_017012852.1 | XP_016868341.1 | 4 | 1130 | |||
| GenBank mRNA | AB014605.2 | - | 4 | 880 | ||
| AF038563.1 | - | 4 | 899 | |||
| AK297353.1 | - | 4 | 884 | |||
| AK297580.1 | - | 3 | 458 | |||
| AK298988.1 | - | 4 | 774 | |||
| BC150277.1 | - | 4 | 880 | 9007255 | ||