Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAGI2 SIRNA Assays ›
          Gene Symbol:
          MAGI2
          Gene Name:
          membrane associated guanylate kinase, WW and PDZ domain containing 2
          Gene Aliases:
          ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM
          Chromosome Location:
          Chr.7: 78017055 - 79453805 on Build GRCh38
          UniGene ID:
          Hs.603842
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19100

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAGI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301128.1 NP_001288057.1 6 1262
          NM_012301.3 NP_036433.2 6 1262
          XM_011516718.1 XP_011515020.1 6 1493
          XM_011516719.2 XP_011515021.1 5 4781
          XM_011516720.2 XP_011515022.1 5 9771
          XM_011516726.2 XP_011515028.1 2 437
          XM_011516728.1 XP_011515030.1 2 102
          XM_017012840.1 XP_016868329.1 6 1493
          XM_017012841.1 XP_016868330.1 6 1493
          XM_017012842.1 XP_016868331.1 6 1493
          XM_017012843.1 XP_016868332.1 6 1493
          XM_017012844.1 XP_016868333.1 6 1493
          XM_017012845.1 XP_016868334.1 6 1493
          XM_017012846.1 XP_016868335.1 6 1493
          XM_017012847.1 XP_016868336.1 6 7730
          XM_017012848.1 XP_016868337.1 5 4778
          XM_017012849.1 XP_016868338.1 5 4778
          XM_017012850.1 XP_016868339.1 6 1493
          XM_017012851.1 XP_016868340.1 6 1493
          XM_017012852.1 XP_016868341.1 6 1493
          GenBank mRNA AK297353.1 - 6 1247
          AK297580.1 - 5 821
          AK298988.1 - 6 1137
          AK308685.1 - 2 83
          AK309012.1 - 2 104

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          planar cell polarity pathway involved in axis elongation
          negative regulation of cell proliferation
          positive regulation of neuron projection development
          negative regulation of cell migration
          positive regulation of phosphoprotein phosphatase activity
          negative regulation of activin receptor signaling pathway
          nerve growth factor signaling pathway
          receptor clustering
          protein heterooligomerization
          negative regulation of protein kinase B signaling
          SMAD protein signal transduction
          mitotic cell cycle arrest
          glomerular visceral epithelial cell development
          neuroligin clustering involved in postsynaptic membrane assembly
          cellular response to nerve growth factor stimulus
          positive regulation of synaptic vesicle clustering
          signal transducer activity
          protein binding
          phosphatase binding
          receptor signaling complex scaffold activity
          beta-1 adrenergic receptor binding
          SMAD binding
          type II activin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lcx2p:80/100.66.77.34:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0