Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198782.2 | NP_942077.2 | 1, 2 | 620 | ||
XM_006236333.2 | XP_006236395.1 | 1, 2 | 442 | |||
GenBank mRNA | AB058962.1 | - | 1, 2 | 620 | ||
AB058963.1 | - | 1, 2 | 620 | |||
AB058964.1 | - | 1, 2 | 620 | |||
AW526027.1 | - | 1, 2 | 76 |