Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001068704.5 | XP_001068704.3 | 9 | 1534 | ||
XM_006249947.2 | XP_006250009.1 | 9 | 1460 | |||
XM_006249949.2 | XP_006250011.1 | 8 | 1357 | |||
XM_006249950.2 | XP_006250012.1 | 7 | 1174 | |||
GenBank mRNA | FQ211198.1 | - | 8 | 1337 |