siRNA Details
Gene Symbol: Klf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127604.1 | NP_001121076.1 | 2 | 1333 | ||
GenBank mRNA | BC166458.1 | - | 2 | 1333 | 9026732 |

More Information
Other Species Targeted

