Search
Search

NAALADL1| Gene Symbol: NAALADL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005468.2 | NP_005459.2 | 13 | 1599 | ||
| XM_011544707.2 | XP_011543009.1 | 14 | 1567 | |||
| XM_011544709.2 | XP_011543011.1 | 13 | 1939 | |||
| XM_011544711.2 | XP_011543013.1 | 12 | 1873 | |||
| XM_011544712.2 | XP_011543014.1 | 12 | 1874 | |||
| GenBank mRNA | AF010141.1 | - | 4 | 329 | ||
| AJ012371.1 | - | 13 | 1599 | |||