Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGLL5 SIRNA Assays ›
          Gene Symbol:
          IGLL5 IGLC1 IGLL1 IGLV3-21
          Gene Name:
          immunoglobulin lambda like polypeptide 5
          immunoglobulin lambda constant 1
          immunoglobulin lambda like polypeptide 1
          immunoglobulin lambda variable 3-21
          Gene Aliases:
          IGL, IGLV, VL-MAR
          IGLC
          14.1, AGM2, CD179b, IGL1, IGL5, IGLJ14.1, IGLL, IGO, IGVPB, VPREB2
          IGLV321, V2-14
          Chromosome Location:
          Chr.22: 22887780 - 22895834 on Build GRCh38
          Chromosome Location:
          Chr.22: 22895375 - 22895694 on Build GRCh38
          Chromosome Location:
          Chr.22: 23573125 - 23580548 on Build GRCh38
          Chromosome Location:
          Chr.22: 22712419 - 22713199 on Build GRCh38
          UniGene ID:
          Hs.449585
          Hs.449585
          Hs.348935
          Hs.449585
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s194557

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IGLL5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178126.1 NP_001171597.1 3 791
          NM_001256296.1 NP_001243225.1 2 672
          GenBank mRNA AB027448.1 - 3 526
          AB064153.1 - 3 589
          AB064158.1 - 3 589
          AB064173.1 - 577
          AB064175.1 - 3 589
          AB064186.1 - 3 586
          AB064202.1 - 589
          AB064207.1 - 3 589
          AB064212.1 - 3 583
          AB064220.1 - 3 589
          AB064233.1 - 3 580
          AJ318021.1 - 3 794
          AJ318022.1 - 3 791
          AJ319670.1 - 2 623
          BC012876.2 - 4 624 4149333
          BC062711.1 - 4 629 30350364
          BC064491.1 - 4 610 4576612
          BC073784.1 - 4 604 4297904
          BC073795.1 - 4 607 6302957
          CR936629.1 - 4 612
          EF589388.1 - 3 523
          EF589482.1 - 3 520
          EF589498.1 - 3 523
          EF589507.1 - 3 514
          EF589514.1 - 3 523
          JF806287.1 - 4 586
          X57819.1 - 4 581
          X57823.1 - 4 580
          Gene Symbol: IGLC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC007782.2 - 4 589
          Gene Symbol: IGLL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020070.3 NP_064455.1 3 632
          NM_152855.2 NP_690594.1 2 516
          XM_011530169.2 XP_011528471.1 3 875
          GenBank mRNA BC012293.1 - 3 572 4508892
          BC030239.1 - 2 458
          M27749.1 - 3 632
          Gene Symbol: IGLV3-21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC070352.1 - 4 614

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagocytosis, recognition
          phagocytosis, engulfment
          complement activation, classical pathway
          defense response to bacterium
          innate immune response
          B cell receptor signaling pathway
          positive regulation of B cell activation
          immune response
          antigen binding
          immunoglobulin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0