Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000207.2 | NP_000198.1 | 3 | 343 | ||
NM_001185097.1 | NP_001172026.1 | 3 | 369 | |||
NM_001185098.1 | NP_001172027.1 | 2 | 522 | |||
NM_001291897.1 | NP_001278826.1 | 3 | 403 | |||
GenBank mRNA | AY899304.1 | - | 3 | 339 | ||
BC005255.1 | - | 3 | 343 | 3950204 | ||
BM510347.1 | - | 3 | 364 | |||
BM510748.1 | - | 3 | 347 | |||
BP322143.1 | - | 2 | 522 | |||
BT006808.1 | - | 2 | 284 | |||
DQ778082.1 | - | 2 | 248 | |||
JF909299.1 | - | 2 | 237 | |||
JQ951950.1 | - | 3 | 328 | |||
X70508.1 | - | 3 | 328 |