Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other S100A9 SIRNA Assays ›
          Gene Symbol:
          S100A9
          Gene Name:
          S100 calcium binding protein A9
          Gene Aliases:
          60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
          Chromosome Location:
          Chr.1: 153357854 - 153361027 on Build GRCh38
          UniGene ID:
          Hs.112405
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s194779

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: S100A9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002965.3 NP_002956.1 2 98
          GenBank mRNA AF086362.1 - 1 79
          AK311882.1 - 2 99
          BC047681.2 - 1 80 6060124
          CR542207.1 - 1 55
          CR542224.1 - 1 55
          M26311.1 - 1 86
          X06233.1 - 2 99

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          leukocyte migration involved in inflammatory response
          positive regulation of peptide secretion
          regulation of translation
          autophagy
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          signal transduction
          cell-cell signaling
          astrocyte development
          positive regulation of blood coagulation
          positive regulation of cell growth
          neutrophil chemotaxis
          actin cytoskeleton reorganization
          sequestering of zinc ion
          chemokine production
          peptidyl-cysteine S-trans-nitrosylation
          defense response to bacterium
          innate immune response
          regulation of integrin biosynthetic process
          positive regulation of inflammatory response
          defense response to fungus
          positive regulation of NF-kappaB transcription factor activity
          regulation of cytoskeleton organization
          neutrophil aggregation
          cellular oxidant detoxification
          positive regulation of intrinsic apoptotic signaling pathway
          signal transducer activity
          calcium ion binding
          protein binding
          microtubule binding
          zinc ion binding
          antioxidant activity
          Toll-like receptor 4 binding
          arachidonic acid binding
          RAGE receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5wt4x:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0