Search
Search

CCL17| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002987.2 | NP_002978.1 | 4 | 379 | ||
| XM_011523256.2 | XP_011521558.1 | 6 | 750 | |||
| XM_017023530.1 | XP_016879019.1 | 6 | 753 | |||
| GenBank mRNA | BC069107.1 | - | 4 | 379 | 7216923 | |
| BC112066.1 | - | 4 | 345 | 8327534 | ||
| BC112068.1 | - | 4 | 348 | 8327536 | ||
| D43767.1 | - | 3 | 302 | |||