Search
Search

| Gene Symbol: HIST2H4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003548.2 | NP_003539.1 | 1 | 94 | ||
| GenBank mRNA | BC019846.1 | - | 1 | 73 | ||
| BC093969.1 | - | 1 | 88 | 7939814 | ||
| BC108260.1 | - | 1 | 83 | |||
| BC112193.1 | - | 1 | 88 | 8327661 | ||
| CR542172.1 | - | 1 | 66 | |||
| Gene Symbol: HIST2H4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034077.4 | NP_001029249.1 | 1 | 94 | ||