Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195011
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204108.1
NP_001191037.1
2
617
NM_001204109.1
NP_001191038.1
2
617
NM_001204111.1
NP_001191040.1
3
437
NM_001204112.1
NP_001191041.1
3
437
NM_001204113.1
NP_001191042.1
3
437
NM_006538.4
NP_006529.1
3
437
NM_138621.4
NP_619527.1
2
617
NM_138622.3
NP_619528.1
2
617
NM_138623.3
NP_619529.1
3
437
NM_138624.3
NP_619530.1
2
617
NM_138626.3
NP_619532.1
2
617
NM_207002.3
NP_996885.1
3
437
XM_005263550.3
XP_005263607.1
2
1395
XM_005263551.2
XP_005263608.1
2
1395
XM_005263552.3
XP_005263609.1
3
1215
XM_005263553.2
XP_005263610.1
3
1215
XM_005263554.2
XP_005263611.1
2
1395
XM_005263556.3
XP_005263613.1
2
617
XM_005263557.4
XP_005263614.1
2
694
XM_005263561.3
XP_005263618.1
3
437
XM_011510461.2
XP_011508763.1
2
1395
XM_011510464.2
XP_011508766.1
3
1215
XM_017003101.1
XP_016858590.1
3
1215
GenBank mRNA
AB071195.1
-
1
329
AB071196.1
-
2
149
AB071197.1
-
1
329
AB071198.1
-
1
329
AF032457.1
-
1
329
AF032458.1
-
2
149
AK290377.1
-
3
437
AK291269.1
-
2
614
AY352518.1
-
2
149
AY423443.1
-
1
329
AY428962.1
-
1
329
BC033694.1
-
2
599
5213713
CN411912.1
-
2
586
DQ849201.1
-
2
149
DQ849202.1
-
2
149
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00197982_m1
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0