Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB3 SIRNA Assays ›
Gene Symbol:
LDB3
Gene Name:
LIM domain binding 3
Gene Aliases:
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location:
Chr.10: 86664785 - 86736072 on Build GRCh38
UniGene ID:
Hs.657271
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195087
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080114.1
NP_001073583.1
2
234
NM_001080115.1
NP_001073584.1
2
234
NM_001080116.1
NP_001073585.1
2
339
NM_001171610.1
NP_001165081.1
3
356
NM_001171611.1
NP_001165082.1
2
234
NM_007078.2
NP_009009.1
2
234
XM_005269464.4
XP_005269521.1
3
403
XM_005269466.4
XP_005269523.1
3
402
XM_005269468.4
XP_005269525.1
3
399
XM_011539184.2
XP_011537486.1
3
404
XM_011539185.2
XP_011537487.1
3
2298
XM_011539186.2
XP_011537488.1
3
404
XM_011539187.2
XP_011537489.1
3
403
XM_011539188.2
XP_011537490.1
3
404
XM_011539189.1
XP_011537491.1
3
298
XM_011539190.1
XP_011537492.1
2
234
XM_011539191.2
XP_011537493.1
3
403
XM_011539193.1
XP_011537495.1
3
298
XM_011539194.1
XP_011537496.1
3
298
XM_011539195.2
XP_011537497.1
3
400
XM_017015606.1
XP_016871095.1
3
2298
XM_017015608.1
XP_016871097.1
3
2298
XM_017015609.1
XP_016871098.1
3
298
GenBank mRNA
AB014513.1
-
2
232
AF276807.1
-
2
211
AF276808.1
-
2
211
AF276809.1
-
2
211
AJ133766.1
-
2
290
AJ133767.1
-
2
211
AJ133768.1
-
2
211
AK294696.1
-
3
291
AK297871.1
-
3
190
AK300692.1
-
3
247
AK304760.1
-
3
298
AL601484.1
-
2
246
BC010929.2
-
2
236
DA898241.1
-
2
339
DC357269.1
-
3
356
KF772970.1
-
2
211
KU178543.1
-
2
211
KU178544.1
-
2
211
KU178545.1
-
2
211
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sarcomere organization
protein kinase C binding
protein binding
cytoskeletal protein binding
zinc ion binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0