Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2A SIRNA Assays ›
Gene Symbol:
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195089
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306158.1
NP_001293087.1
4
424
NM_001306159.1
NP_001293088.1
4
424
NM_001306160.1
NP_001293089.1
3
252
NM_001306161.1
NP_001293090.1
4
428
NM_007082.4
NP_009013.1
4
424
NM_013412.3
NP_038198.1
4
424
XM_005263569.3
XP_005263626.1
4
435
XM_005263570.3
XP_005263627.1
4
402
XM_006712207.3
XP_006712270.1
4
504
XM_006712208.3
XP_006712271.1
4
393
XM_006712209.3
XP_006712272.1
4
547
XM_006712215.3
XP_006712278.1
5
634
XM_011510509.2
XP_011508811.1
4
395
XM_017003203.1
XP_016858692.1
3
1144
XM_017003204.1
XP_016858693.1
4
1084
XM_017003205.1
XP_016858694.1
3
1144
XM_017003206.1
XP_016858695.1
5
545
XM_017003207.1
XP_016858696.1
4
458
XM_017003208.1
XP_016858697.1
4
418
XM_017003209.1
XP_016858698.1
4
402
XM_017003210.1
XP_016858699.1
3
1144
XM_017003211.1
XP_016858700.1
4
435
XM_017003212.1
XP_016858701.1
3
1144
XM_017003213.1
XP_016858702.1
3
1144
XM_017003214.1
XP_016858703.1
4
1084
XM_017003215.1
XP_016858704.1
3
1144
XM_017003216.1
XP_016858705.1
4
547
XM_017003217.1
XP_016858706.1
4
431
XM_017003218.1
XP_016858707.1
5
585
XM_017003219.1
XP_016858708.1
4
498
XM_017003220.1
XP_016858709.1
3
1144
XM_017003221.1
XP_016858710.1
4
420
XM_017003222.1
XP_016858711.1
5
575
XM_017003223.1
XP_016858712.1
3
1144
XM_017003224.1
XP_016858713.1
4
411
XM_017003225.1
XP_016858714.1
4
402
XM_017003226.1
XP_016858715.1
3
1144
XM_017003227.1
XP_016858716.1
4
411
XM_017003228.1
XP_016858717.1
5
539
XM_017003229.1
XP_016858718.1
4
388
XM_017003230.1
XP_016858719.1
4
402
XM_017003231.1
XP_016858720.1
4
439
XM_017003232.1
XP_016858721.1
4
382
XM_017003233.1
XP_016858722.1
3
1144
XM_017003234.1
XP_016858723.1
3
1494
XM_017003235.1
XP_016858724.1
3
1144
XM_017003236.1
XP_016858725.1
4
416
GenBank mRNA
AF095350.1
-
4
352
AF095351.1
-
4
352
AK289496.1
-
5
339
BC111008.1
-
4
418
5769737
BC126129.1
-
3
266
8991845
BC144050.1
-
3
266
9052570
BC144052.1
-
3
266
9052572
BI552699.1
-
4
431
CR456554.1
-
3
259
HY023006.1
-
4
381
HY075940.1
-
4
417
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0