Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 642 | ||
GenBank mRNA | AJ457795.1 | - | 2 | 344 | ||
BC005382.1 | - | 2 | 401 | 3935644 | ||
BC062672.1 | - | 2 | 582 | 4047937 | ||
BC103961.1 | - | 2 | 341 | 40013988 | ||
BC121070.2 | - | 2 | 351 | 40119569 | ||
BC134418.1 | - | 2 | 351 | 40119568 | ||
BM554791.1 | - | 2 | 642 |
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 368 | ||
GenBank mRNA | AF098306.1 | - | 2 | 344 | ||
AJ238277.1 | - | 2 | 347 | |||
AK225988.1 | - | 2 | 367 | |||
BC054023.1 | - | 2 | 369 | 6648369 | ||
BC069393.1 | - | 2 | 344 | 7262682 | ||
BC069816.1 | - | 2 | 344 | 7262670 | ||
CB115288.1 | - | 2 | 368 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 368 | ||
GenBank mRNA | CB110020.1 | - | 2 | 368 | ||
CB113891.1 | - | 2 | 368 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 401 | ||
GenBank mRNA | AA412605.1 | - | 2 | 364 |