Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017914.3 | NP_060384.3 | 3 | 850 | ||
GenBank mRNA | AK000647.1 | - | 3 | 623 | ||
AK096105.1 | - | 2 | 1630 | |||
AK225502.1 | - | 3 | 623 | |||
BC000890.1 | - | 3 | 819 |