Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024326.3 | NP_077302.3 | 4 | 2335 | ||
XM_005270149.2 | XP_005270206.1 | 6 | 1679 | |||
XM_005270150.4 | XP_005270207.1 | 5 | 2129 | |||
XM_005270151.3 | XP_005270208.1 | 5 | 1285 | |||
XM_017016631.1 | XP_016872120.1 | 5 | 1416 | |||
GenBank mRNA | BC002912.2 | - | 4 | 1152 | 3944940 | |
BC036120.1 | - | 4 | 2316 | 5286464 |