siRNA Details
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030789.3 | NP_110416.1 | 2 | 414 | ||
NM_178580.2 | NP_848695.1 | 2 | 414 | |||
NM_178581.2 | NP_848696.1 | 2 | 414 | |||
NM_178582.2 | NP_848697.1 | 2 | 414 | |||
GenBank mRNA | AF172086.1 | - | 2 | 365 | ||
AF483215.1 | - | 2 | 360 | |||
AF515663.1 | - | 2 | 272 | |||
AJ345029.1 | - | 2 | 357 | |||
AJ420895.1 | - | 2 | 296 | |||
AK074686.1 | - | 2 | 339 | |||
AK075283.1 | - | 2 | 374 | |||
AK314410.1 | - | 2 | 382 | |||
AY169310.1 | - | 2 | 371 | |||
AY169311.1 | - | 2 | 371 | |||
AY169312.1 | - | 2 | 371 | |||
BC008938.2 | - | 2 | 385 | 2961249 | ||
BC008959.2 | - | 2 | 385 | 2961249 | ||
BC062595.1 | - | 2 | 365 | 5245982 | ||
BI192962.1 | - | 2 | 386 | |||
BI552780.1 | - | 2 | 399 | |||
DA087240.1 | - | 2 | 414 | |||
DQ168450.1 | - | 2 | 366 | |||
KU178790.1 | - | 2 | 272 | |||
KU178791.1 | - | 2 | 272 |

More Information
Other Species Targeted

