Search
Search

KMT5C| Gene Symbol: KMT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032701.3 | NP_116090.2 | 3, 4 | 525 | ||
| XM_011527415.2 | XP_011525717.1 | 3, 4 | 1045 | |||
| XM_017027388.1 | XP_016882877.1 | 3, 4 | 498 | |||
| GenBank mRNA | AF289582.1 | - | 2, 3 | 598 | ||
| AK308586.1 | - | 3, 4 | 525 | |||
| BC044889.1 | - | 3, 4 | 482 | 3461265 | ||