Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM86B1 SIRNA Assays ›
          Gene Symbol:
          FAM86B1 FAM86B2
          Gene Name:
          family with sequence similarity 86 member B1
          family with sequence similarity 86 member B2
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 12182104 - 12194118 on Build GRCh38
          Chromosome Location:
          Chr.8: 12424419 - 12436418 on Build GRCh38
          UniGene ID:
          Hs.675799
          Hs.458413
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s195505

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM86B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083537.1 NP_001077006.1 7 1493
          NR_003494.1 6 1227
          XM_006716256.3 XP_006716319.1 8 1588
          XM_006716257.3 XP_006716320.1 7 1658
          XM_006716260.3 XP_006716323.1 6 1402
          XM_006716262.1 XP_006716325.1 6 1245
          XM_011543842.2 XP_011542144.1 6 1667
          XM_011543843.2 XP_011542145.1 7 1606
          XM_017013916.1 XP_016869405.1 7 1634
          XM_017013917.1 XP_016869406.1 6 1555
          XM_017013918.1 XP_016869407.1 6 1527
          XM_017013919.1 XP_016869408.1 5 1116
          GenBank mRNA AK098138.1 - 7 1090
          BC062615.1 - 7 1167
          BC085016.1 - 8 1196
          BC093808.1 - 1 42
          BC093810.1 - 1 42
          BC098427.1 - 7 1173
          Gene Symbol: FAM86B2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005273603.4 XP_005273660.1 7 1523
          XM_005273604.4 XP_005273661.1 6 1442
          XM_005273605.4 XP_005273662.1 6 1343
          XM_005273607.2 XP_005273664.1 6 1381
          XM_006716387.3 XP_006716450.1 7 1544
          XM_011544618.2 XP_011542920.1 8 1595
          XM_011544619.2 XP_011542921.1 7 1562
          XM_011544622.2 XP_011542924.1 7 1464
          XM_017013740.1 XP_016869229.1 6 1525
          XM_017013741.1 XP_016869230.1 5 1462
          XM_017013742.1 XP_016869231.1 5 1444

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          methylation
          methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-l2qgx:80/100.66.79.25:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0