Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR1H3 SIRNA Assays ›
          Gene Symbol:
          NR1H3
          Gene Name:
          nuclear receptor subfamily 1 group H member 3
          Gene Aliases:
          LXR-a, LXRA, RLD-1
          Chromosome Location:
          Chr.11: 47248300 - 47269033 on Build GRCh38
          UniGene ID:
          Hs.438863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19570

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR1H3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130101.2 NP_001123573.1 3, 4 487
          NM_001130102.2 NP_001123574.1 2, 3 296
          NM_001251934.1 NP_001238863.1 3, 4 435
          NM_001251935.1 NP_001238864.1 3, 4 476
          NM_005693.3 NP_005684.2 3, 4 487
          XM_005252705.1 XP_005252762.1 4, 5 440
          XM_005252706.1 XP_005252763.1 4, 5 557
          XM_005252707.4 XP_005252764.1 3, 4 573
          XM_005252709.1 XP_005252766.1 4, 5 441
          XM_005252710.1 XP_005252767.1 3, 4 339
          XM_005252713.3 XP_005252770.1 3, 4 362
          XM_006718112.1 XP_006718175.1 4, 5 429
          XM_006718113.1 XP_006718176.1 4, 5 440
          XM_006718115.1 XP_006718178.1 3, 4 447
          XM_006718116.1 XP_006718179.1 3, 4 356
          XM_011519805.2 XP_011518107.1 3, 4 359
          XM_011519806.1 XP_011518108.1 2, 3 323
          XM_017017056.1 XP_016872545.1 2, 3 323
          XM_017017057.1 XP_016872546.1 2, 3 323
          XM_017017058.1 XP_016872547.1 2, 3 272
          GenBank mRNA AB307698.1 - 2, 3 237
          AK122661.1 - 2, 3 1475
          AK290614.1 - 3, 4 442
          AK302132.1 - 3, 4 424
          AK309027.1 - 3, 4 427
          AK311590.1 - 3, 4 476
          AK313311.1 - 3, 4 435
          AU135617.1 - 2, 3 296
          BC008819.2 - 3, 4 406 3957848
          BC041172.1 - 2, 3 278 5744574
          BM835230.1 - 2, 3 296
          BP300071.1 - 3, 4 458
          CV026805.1 - 2, 3 237
          HQ692819.1 - 2, 3 237
          U22662.1 - 2, 3 272

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          negative regulation of macrophage derived foam cell differentiation
          positive regulation of triglyceride biosynthetic process
          positive regulation of receptor biosynthetic process
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          intracellular receptor signaling pathway
          positive regulation of cellular protein metabolic process
          negative regulation of lipid transport
          positive regulation of cholesterol transport
          response to progesterone
          positive regulation of toll-like receptor 4 signaling pathway
          cholesterol homeostasis
          regulation of circadian rhythm
          negative regulation of macrophage activation
          apoptotic cell clearance
          steroid hormone mediated signaling pathway
          positive regulation of fatty acid biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of pinocytosis
          negative regulation of inflammatory response
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          sterol homeostasis
          negative regulation of interferon-gamma-mediated signaling pathway
          triglyceride homeostasis
          cellular response to lipopolysaccharide
          negative regulation of pancreatic juice secretion
          negative regulation of secretion of lysosomal enzymes
          regulation of cholesterol homeostasis
          positive regulation of cholesterol homeostasis
          DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          zinc ion binding
          cholesterol binding
          sterol response element binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dhnrf:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0