Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320778.1 | NP_001307707.1 | 6 | 651 | ||
NM_005697.4 | NP_005688.2 | 5 | 522 | |||
XM_017021856.1 | XP_016877345.1 | 6 | 576 | |||
GenBank mRNA | AF005038.2 | - | 5 | 427 | ||
AK096376.1 | - | 4 | 413 | |||
AK096794.1 | - | 4 | 2311 | |||
AK291884.1 | - | 5 | 508 | |||
AK315516.1 | - | 5 | 471 | |||
BC001376.2 | - | 5 | 472 | 3050527 | ||
BC004385.1 | - | 5 | 472 | 3617954 | ||
CD300387.1 | - | 6 | 627 | |||
CR456800.1 | - | 5 | 398 |