Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320724.1 | NP_001307653.1 | 3 | 473 | ||
NM_016940.2 | NP_058636.1 | 2 | 368 | |||
GenBank mRNA | AF212232.1 | - | 2 | 329 | ||
AK000056.1 | - | 2 | 372 | |||
AK223357.1 | - | 2 | 379 | |||
BC012546.1 | - | 2 | 336 | 4291536 | ||
BC017912.1 | - | 2 | 354 | 4291966 | ||
DB039085.1 | - | 3 | 470 | |||
Y19009.1 | - | 2 | 347 |