Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HUWE1 SIRNA Assays ›
Gene Symbol:
HUWE1
Gene Name:
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases:
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location:
Chr.X: 53532096 - 53686729 on Build GRCh38
UniGene ID:
Hs.136905
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19595
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HUWE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031407.6
NP_113584.3
5, 6
546
XM_005261965.3
XP_005262022.1
4, 5
613
XM_011530747.1
XP_011529049.1
5, 6
540
XM_011530751.2
XP_011529053.1
4, 5
408
XM_017029191.1
XP_016884680.1
3, 4
3271
XM_017029192.1
XP_016884681.1
3, 4
3271
XM_017029193.1
XP_016884682.1
3, 4
3271
XM_017029194.1
XP_016884683.1
3, 4
3271
XM_017029195.1
XP_016884684.1
3, 4
3271
XM_017029196.1
XP_016884685.1
3, 4
3271
XM_017029197.1
XP_016884686.1
3, 4
3271
XM_017029198.1
XP_016884687.1
4, 5
613
XM_017029199.1
XP_016884688.1
5, 6
540
XM_017029200.1
XP_016884689.1
4, 5
410
XM_017029201.1
XP_016884690.1
4, 5
427
XM_017029202.1
XP_016884691.1
4, 5
403
XM_017029203.1
XP_016884692.1
4, 5
409
XM_017029204.1
XP_016884693.1
3, 4
3270
XM_017029205.1
XP_016884694.1
3, 4
3271
XM_017029206.1
XP_016884695.1
3, 4
3271
GenBank mRNA
AY772009.1
-
3, 4
191
AY929612.1
-
2, 3
143
BC018750.1
-
5, 6
545
BC038184.1
-
5, 6
545
DQ097177.1
-
5, 6
545
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
base-excision repair
protein monoubiquitination
histone ubiquitination
cell differentiation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
DNA binding
ubiquitin-protein transferase activity
protein binding
ligase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0