Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSPAN32 SIRNA Assays ›
          Gene Symbol:
          TSPAN32
          Gene Name:
          tetraspanin 32
          Gene Aliases:
          ART1, PHEMX, PHMX, TSSC6
          Chromosome Location:
          Chr.11: 2299480 - 2318201 on Build GRCh38
          UniGene ID:
          Hs.271954
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19603

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSPAN32
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_139022.2 NP_620591.3 5 522
          XM_005252719.4 XP_005252776.1 5 516
          XM_005252720.4 XP_005252777.1 5 516
          XM_011519815.2 XP_011518117.2 4 438
          XM_011519818.2 XP_011518120.2 4 438
          XM_011519819.1 XP_011518121.1 5 404
          XM_011519822.1 XP_011518124.1 5 516
          XM_011519828.2 XP_011518130.1 4 313
          XM_011519829.2 XP_011518131.2 5 513
          XM_017017064.1 XP_016872553.1 4 988
          XM_017017065.1 XP_016872554.1 4 988
          XM_017017066.1 XP_016872555.1 4 988
          XM_017017067.1 XP_016872556.1 5 408
          XM_017017068.1 XP_016872557.1 4 988
          XM_017017069.1 XP_016872558.1 4 988
          XM_017017070.1 XP_016872559.1 4 988
          XM_017017071.1 XP_016872560.1 4 988
          GenBank mRNA AF125569.1 - 4 440
          AF176070.1 - 4 455
          AF176071.1 - 4 290
          AK128812.1 - 6 594
          AK298692.1 - 5 348
          AK307638.1 - 6 594
          AK310116.1 - 4 313
          AY039001.1 - 5 391
          AY303780.1 - 3 220
          BC016693.1 - 5 526 4071111
          BG506933.1 - 5 522

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          cell surface receptor signaling pathway
          integrin-mediated signaling pathway
          cell-cell signaling
          negative regulation of cell proliferation
          negative regulation of myeloid dendritic cell activation
          defense response to protozoan
          regulation of defense response to virus
          platelet aggregation
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0