Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1964
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
815
NM_001143806.1
NP_001137278.1
2
1030
NM_001143807.1
NP_001137279.1
2
916
NM_001143808.1
NP_001137280.1
2
752
NM_001143809.1
NP_001137281.1
2
762
NM_001143810.1
NP_001137282.1
3
1045
NM_001143811.1
NP_001137283.1
4
1162
NM_001143812.1
NP_001137284.1
2
904
NM_001143813.1
NP_001137285.1
2
986
NM_001143814.1
NP_001137286.1
3
1111
NM_001143816.1
NP_001137288.1
2
1509
NM_001709.4
NP_001700.2
2
1004
NM_170731.4
NP_733927.1
2
1326
NM_170732.4
NP_733928.1
2
1113
NM_170733.3
NP_733929.1
2
1016
NM_170734.3
NP_733930.1
2
964
NM_170735.5
NP_733931.1
1
1743
XM_011520280.2
XP_011518582.1
2
1910
GenBank mRNA
AF400438.1
-
1
676
AK289763.1
-
2
812
AK289853.1
-
2
713
AY054392.2
-
2
1017
AY054393.2
-
2
759
AY054394.2
-
2
974
AY054395.2
-
2
1057
AY054396.2
-
2
815
AY054397.2
-
2
898
AY054398.2
-
2
945
AY054399.2
-
3
830
AY054400.2
-
2
931
AY054406.3
-
3
655
AY656701.1
-
1
658
BC029795.1
-
2
989
5193877
EF674517.1
-
2
815
EF674518.1
-
2
1030
EF674519.1
-
2
1113
EF674520.1
-
2
916
EF674521.1
-
2
1016
EF689009.1
-
1
1757
EF689010.1
-
2
1523
EF689011.1
-
2
752
EF689012.1
-
2
762
EF689013.1
-
2
904
EF689014.1
-
2
986
EF689015.1
-
2
1004
EF689016.1
-
3
1111
EF689017.1
-
3
863
EF689018.1
-
2
964
EF689019.1
-
3
1045
EF689020.1
-
4
1162
EF689021.1
-
2
1326
KC855559.1
-
1
194
KP229412.1
-
1
658
KU177943.1
-
1
658
KU177944.1
-
2
541
KU177945.1
-
2
514
M61176.1
-
1
1220
X60201.1
-
2
954
X91251.1
-
2
942
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: AY176065.
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-tfbk7:80/10.239.166.103:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0