Search
Search

CFB| Gene Symbol: CFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001710.5 | NP_001701.2 | 2 | 538 | ||
| GenBank mRNA | AF349679.1 | - | 2 | 260 | ||
| AK130533.1 | - | 2 | 467 | |||
| AK223400.1 | - | 2 | 386 | |||
| AK304042.1 | - | 2 | 507 | |||
| BC004143.2 | - | 2 | 395 | 2959705 | ||
| BC007990.2 | - | 2 | 395 | 2959706 | ||
| CD689714.1 | - | 2 | 405 | |||
| L15702.1 | - | 2 | 300 | |||
| S67310.1 | - | 2 | 371 | |||
| X00284.1 | - | 2 | 215 | |||
| X72875.1 | - | 2 | 260 | |||