Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320623.1 | NP_001307552.1 | 2 | 792 | ||
NM_001320627.1 | NP_001307556.1 | 2 | 792 | |||
NM_001320628.1 | NP_001307557.1 | 1 | 110 | |||
NR_135316.1 | 2 | 792 | ||||
GenBank mRNA | AF271775.1 | - | 2 | 387 | ||
AK027255.1 | - | 2 | 185 | |||
AK295153.1 | - | 2 | 96 | |||
BC062756.1 | - | 2 | 173 | |||
BC130401.1 | - | 2 | 479 | 40146432 | ||
BC130403.1 | - | 2 | 479 | 40146434 | ||
BI517472.1 | - | 2 | 142 | |||
BM696373.1 | - | 1 | 95 | |||
DA438012.1 | - | 2 | 184 |
Gene Symbol: BOLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031827.2 | NP_001026997.1 | 2 | 792 | ||
NR_135304.1 | 1 | 110 | ||||
GenBank mRNA | AF060511.2 | - | 2 | 181 | ||
AK307153.1 | - | 2 | 184 | |||
BC022832.1 | - | 2 | 792 | 4647797 | ||
BC144688.1 | - | 2 | 479 | 9053222 | ||
BQ639539.1 | - | 1 | 87 | |||
BU590412.1 | - | 2 | 49 |
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039182.2 | NP_001034271.1 | 2 | 792 | ||
GenBank mRNA | BC137532.1 | - | 2 | 479 | 9021532 | |
BC137534.1 | - | 2 | 479 | 9021534 |