Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEACAM1 SIRNA Assays ›
          Gene Symbol:
          CEACAM1
          Gene Name:
          carcinoembryonic antigen related cell adhesion molecule 1
          Gene Aliases:
          BGP, BGP1, BGPI
          Chromosome Location:
          Chr.19: 42507306 - 42528509 on Build GRCh38
          UniGene ID:
          Hs.512682
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1977

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEACAM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024912.2 NP_001020083.1 5 1343
          NM_001205344.1 NP_001192273.1 5 1343
          NM_001712.4 NP_001703.2 5 1343
          GenBank mRNA AK314211.1 - 5 1315
          AL833584.1 - 5 1313
          BC014473.1 - 5 1300 4871488
          BC024164.1 - 5 1296
          D90311.1 - 5 1272
          D90313.1 - 5 1149
          J03858.1 - 5 1289
          M72238.1 - 5 1303
          S71326.1 - 5 1208
          X16354.1 - 5 1280
          X16356.1 - 4 936

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          regulation of cell growth
          blood vessel development
          negative regulation of T cell mediated cytotoxicity
          negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          negative regulation of protein kinase activity
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          integrin-mediated signaling pathway
          regulation of endothelial cell migration
          regulation of phosphatidylinositol 3-kinase signaling
          bile acid and bile salt transport
          cell migration
          regulation of cell migration
          negative regulation of granulocyte differentiation
          negative regulation of interleukin-1 production
          cellular response to insulin stimulus
          common myeloid progenitor cell proliferation
          insulin receptor internalization
          granulocyte colony-stimulating factor signaling pathway
          regulation of epidermal growth factor receptor signaling pathway
          negative regulation of vascular permeability
          negative regulation of cytotoxic T cell degranulation
          wound healing, spreading of cells
          regulation of endothelial cell differentiation
          negative regulation of fatty acid biosynthetic process
          negative regulation of T cell receptor signaling pathway
          leukocyte migration
          negative regulation of lipid biosynthetic process
          regulation of blood vessel remodeling
          regulation of ERK1 and ERK2 cascade
          negative regulation of platelet aggregation
          cell-cell adhesion
          insulin catabolic process
          regulation of homophilic cell adhesion
          regulation of sprouting angiogenesis
          negative regulation of hepatocyte proliferation
          positive regulation of vasculogenesis
          molecular_function
          actin binding
          protein binding
          bile acid transmembrane transporter activity
          kinase binding
          protein phosphatase binding
          filamin binding
          protein homodimerization activity
          protein dimerization activity
          protein tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0