Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144026.2 | NP_001137498.1 | 3, 4 | 422 | ||
NM_001144027.2 | NP_001137499.1 | 3, 4 | 422 | |||
NM_001144028.2 | NP_001137500.1 | 3, 4 | 422 | |||
NM_014434.3 | NP_055249.1 | 3, 4 | 422 | |||
XM_011518545.1 | XP_011516847.1 | 3, 4 | 412 | |||
XM_011518547.1 | XP_011516849.1 | 3, 4 | 212 | |||
XM_017014626.1 | XP_016870115.1 | 3, 4 | 422 | |||
XM_017014627.1 | XP_016870116.1 | 3, 4 | 422 | |||
GenBank mRNA | AF199509.1 | - | 3, 4 | 358 | ||
AK026089.1 | - | 3, 4 | 394 | |||
AK074403.1 | - | 3, 4 | 383 | |||
AK225995.1 | - | 3, 4 | 393 | |||
AK290026.1 | - | 3, 4 | 419 | |||
AY077845.1 | - | 3, 4 | 340 | |||
BC015735.1 | - | 3, 4 | 358 | 4869561 | ||
BC078668.1 | - | 3, 4 | 358 | |||
BC093782.1 | - | 3, 4 | 339 | 7939627 | ||
BC111943.1 | - | 3, 4 | 339 | 8327411 | ||
DA408370.1 | - | 3, 4 | 422 | |||
DA858250.1 | - | 3, 4 | 422 |